Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) nitrate (0.2% Pd) in nitric acid 5% matrix modifier solution

Supplier: Ricca Chemical
Danger

RMMPD2KN-500N RMMPD2KN-100N RMMPD2KN-50N
RCMMPD2KN-50EA 282.16 USD InStock
RCMMPD2KN-50 RCMMPD2KN-500 RCMMPD2KN-100
Palladium(II) nitrate (0.2% Pd) in nitric acid 5% matrix modifier solution
Palladium (II) nitrate


Formula: N₂O₆Pd
MW: 230.43 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, brown, odorless
Certified Concentration 2000 ± 10 ppm Pd

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty