Would you like to log in again?
Zirconium ICP-MS Standard, 1 ml = 1 mg Zr (1000 ppm Zr)ZrO(NO₃)₂ in 3% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
RMSZR1KN-100N
RCMSZR1KN-100EA 562.03 USD InStock
RCMSZR1KN-100
Zirconium ICP-MS Standard, 1 ml = 1 mg Zr (1000 ppm Zr)ZrO(NO₃)₂ in 3% HNO₃, RICCA Chemical Company
Standards Cationic Standards Zirconium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty