Would you like to log in again?
Calcium ICP Standard, 1 ml = 1 mg Ca (1,000 ppm Ca)CaCO₃ in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RPCA1KN0-100N RPCA1KN0-500N
RCPCA1KN-500EA 283.82 USD InStock
RCPCA1KN-500 RCPCA1KN-100
Calcium ICP Standard, 1 ml = 1 mg Ca (1,000 ppm Ca)CaCO₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Calcium Standards
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty