Would you like to log in again?
Cesium ICP Standard, 1 ml = 10 mg Cs (10,000 ppm Cs)CsNO₃ in 5% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPCS10KN-100N RPCS10KN-500N
RCPCS10KN-100EA 305.25 USD InStock
RCPCS10KN-100 RCPCS10KN-500
Cesium ICP Standard, 1 ml = 10 mg Cs (10,000 ppm Cs)CsNO₃ in 5% HNO₃, Ricca Chemical Company
Standards Cationic Standards Cesium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty