Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 5% (v/v) acid blank

Supplier: Ricca Chemical
Warning

RPHACID5-500N
RCPHACID5-500EA 120.21 USD InStock
RCPHACID5-500
Hydrochloric acid 5% (v/v) acid blank
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.02…1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Aluminum (Al) Actual Value Reported
Antimony (Sb) Actual Value Reported
Appearance Clear, colorless, odorless
Arsenic (As) Actual Value Reported
Barium (Ba) Actual Value Reported
Beryllium (Be) Actual Value Reported
Bismuth (Bi) Actual Value Reported
Boron (B) Actual Value Reported
Cadmium (Cd) Actual Value Reported
Calcium (Ca) Actual Value Reported

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty