Would you like to log in again?
Neodymium ICP Standard, 1 mL = 1 mg Nd (1,000 ppm Nd)Nd₂O₃ in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPND1KN0-100N
RCPND1KN-100EA 145.36 USD InStock
RCPND1KN-100
Neodymium ICP Standard, 1 mL = 1 mg Nd (1,000 ppm Nd)Nd₂O₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Neodymium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty