Would you like to log in again?
Nickel ICP Standard, 1 ml = 1 mg Ni (1000 ppm Ni)Ni in 3% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
RPNI1KN0-100N RPNI1KN0-500N
RCPNI1KN-500EA 305.27 USD InStock
RCPNI1KN-500 RCPNI1KN-100
Nickel ICP Standard, 1 ml = 1 mg Ni (1000 ppm Ni)Ni in 3% HNO₃, RICCA Chemical Company
Standards Cationic Standards Nickel Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty