Would you like to log in again?
Phosphorus ICP Standard, 1 ml = 10 mg P (10000 ppm P) NH₄H₂PO₄ in H₂O, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPP10KW0-50N RPP10KW0-100N RPP10KW0-500N
RCPP10KW-50EA 142.31 USD InStock
RCPP10KW-50 RCPP10KW-500 RCPP10KW-100
Phosphorus ICP Standard, 1 ml = 10 mg P (10000 ppm P) NH₄H₂PO₄ in H₂O, Ricca Chemical Company
Standards Cationic Standards Phosphorus Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty