Would you like to log in again?
Palladium ICP Standard, 1 ml = 1 mg Pd (1000 ppm Pd)Pd in 3% HCl, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPPD1KH0-500N RPPD1KH0-100N
RCPPD1KH-500EA 699.52 USD InStock
RCPPD1KH-500 RCPPD1KH-100
Palladium ICP Standard, 1 ml = 1 mg Pd (1000 ppm Pd)Pd in 3% HCl, Ricca Chemical Company
Standards Cationic Standards Palladium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty