Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Strontium ICP Standard, 1 ml = 1 mg Sr (1,000 ppm Sr)Sr(NO₃)₂ in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPSR1KN0-500N RPSR1KN0-100N
RCPSR1KN-100EA 137.84 USD InStock
RCPSR1KN-100 RCPSR1KN-500
Strontium ICP Standard, 1 ml = 1 mg Sr (1,000 ppm Sr)Sr(NO₃)₂ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Strontium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty