Would you like to log in again?
Titanium ICP Standard, 1 ml = 10 mg Ti (10,000 ppm Ti)(NH₄)2TiF₆ in H₂O / trace HF, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPTI10KW-50N RPTI10KW-100N
RCPTI10KW-100EA 254.1 USD InStock
RCPTI10KW-100 RCPTI10KW-50
Titanium ICP Standard, 1 ml = 10 mg Ti (10,000 ppm Ti)(NH₄)2TiF₆ in H₂O / trace HF, Ricca Chemical Company
Standards Cationic Standards Titanium Standards
10,000 ppm = 10000 µg/mL
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty