Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia USP Test Solution (TS) 2

Supplier: Ricca Chemical
Danger

R0616200-500A
RCR0616200500AEA 203 USD InStock
RCR0616200500A
Ammonia USP Test Solution (TS) 2
Ammonia


Formula: NH₃
MW: 17.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Red) 13.0-14.0 % (v/v)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty