Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia-Cyanide USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R0623300-120B
RCR0623300120BEA 69.4 USD InStock
RCR0623300120B
Ammonia-Cyanide USP Test Solution (TS)
Ammonia-Cyanide


Storage Temperature: Ambient
UN: 2922
ADR: 8 (6.1),III
Order Now
Specification Test Results
Appearance Colorless liquid

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty