Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arsenic(III) oxide solution USP 211

Supplier: Ricca Chemical
Synonyms: di-Arsenic trioxide

R0817000-500A
RCR0817000500AEA 249.36 USD InStock
RCR0817000500A
Arsenic(III) oxide solution USP 211
Arsenic (III) oxide


Formula: As₂O₃
MW: 197.84 g/mol
MDL Number: MFCD00003433
CAS Number: 1327-53-3
UN: 1561
ADR: 6.1,II
Order Now
Specification Test Results
Assay (As) 100 ± 0.5 ppm
Appearance Colorless liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty