Would you like to log in again?
Buffer Concentrate, 5x, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), RICCA Chemical Company
Supplier: RICCA Chemical Company
R1550510-20F
RCR1550510-20FEA 613.67 USD InStock
RCR1550510-20F
Buffer Concentrate, 5x, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), RICCA Chemical Company
Buffers pH Calibration Buffers
5x Concentration. Ideal for limited storage space.
  • Tested in ISO 17025 accredited laboratories
Certifications: ISO 17025
Packaging: 5x Concentrate: To be diluted with 4 parts water before use
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty