Would you like to log in again?
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: RICCA Chemical Company
R1498000-1A R1550000-4A R1489000-20F R1490000-4A 1485-32 R1495000-1A R1540000-4A R1492000-4A R1490000-500A R1498000-500A R1498000-4A R1495000-4A R1492000-1A R1505000-4A R1550000-10F R1565000-1A R1565000-4A R1493000-4A R1495000-20PK R1550000-4F R1540000-500A R1540000-20F R1495000-10F R1540000-1A 1485-16 R1492000-10F R1550000-1A R1510000-10F R1505000-1A R1510000-4A R1498000-20F R1489000-1A 1563-16 R1500000-4F R1489000-500A R1540000-10F R1500000-500A R1493000-500A R1565000-20F R1565000-500A 1485-1 R1500000-4A R1492000-20PK R1492000-500A R1493000-20F R1492000-20F R1493000-10F R1550000-500A R1493000-20PK R1490000-1A R1505000-10F R1500000-20F R1550000-20F R1489000-4A R1495000-20F R1493000-1A R1505000-500A R1495000-500A R1510000-1A R1500000-10F R1510000-500A R1500000-1A R1505000-20F
RC1495-16CS 1389.69 USD InStock
RC1495-16 RC1485-1 RCR1540000-20F RC1489-16 RC1489-1 RC1540-32 RCR1565000-20F RC155016 RC1510-1 RC15505 RC1492-32 RC15501 RC1493-5 RC1490-32 RC1495-1 RC15001 RC1493-1 RC1505-32 RC1563-16 RC1495-5 RCR1565000-1A RCR1489000-20F RC155025 RCR149500020PK RCR1498000-20F RCR1510000-10F RC154016 RC1485-16 RC1540-1 RCR1505000-10F RC150016 RC1510-16 RC1493-32 RCR149300020PK RCR1495000-10F RC1500-1CT RCR1493000-10F RC1489-32 RC1493-16 RCR1540000-10F RC155032 RC1485-32 RCR1565000-4A RC1550-1CT RC150025 RC1498-32 RC15005 RC1498-1 RC1505-16 RC1492-5 RC149216 RC149016 RC1492-1 RCR1565000500A RC149816 RC150032 RCR1505000-20F RCR149200020PK RC1490-1 RC1510-32 RC1505-1 RC1495-32 RCR1492000-10F
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers pH Calibration Buffers
Colorless pH buffer solutions are certified to ±0.01 pH at 25°C
  • Range of packaging is available
Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.
Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty