Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromic acid 10% (w/v)

Supplier: Ricca Chemical
Danger

R2077000-500C R2077000-4C
RCR2077000-4CEA 275.26 USD InStock
RCR2077000-4C RCR2077000500C
Chromic acid 10% (w/v)
Chromic acid


Formula: CrH₂O₄
MW: 118.01 g/mol
Storage Temperature: Ambient
CAS Number: 7738-94-5
UN: 1755
ADR: 8,II
Order Now
Specification Test Results
Appearance Orange liquid
Assay (vs. Sodium Thiosulfate/Starch) 9.9-10.1 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty