Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pararosaniline solution R Reag. Ph. Eur. 1062201, decolorized

Supplier: Ricca Chemical
Danger
Synonyms: Paramagentap-RosanilinePararosaniline baseParafuchsinTris(4-aminophenyl)methanol

R2545500-500C
RCR2545500500CEA 170.91 USD InStock
RCR2545500500C
Pararosaniline solution R Reag. Ph. Eur. 1062201, decolorized
Pararosaniline


Formula: C₁₉H₁₉N₃O
MW: 305.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036222
CAS Number: 467-62-9
Order Now
Specification Test Results
Appearance Colorless to slightly pink liquid
Sensitivity to Aldehyde To Pass Test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty