Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA (Sodium edetate) 0.100 M (M/10) Reag. Ph. Eur. 3005900 volumetric solution

Supplier: Ricca Chemical

R2730100-1A
RCR2730100-1AEA 174.1 USD InStock
RCR2730100-1A
EDTA (Sodium edetate) 0.100 M (M/10) Reag. Ph. Eur. 3005900 volumetric solution
EDTA (Sodium edetate)


Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Calcium Carbonate/Eriochrome Black T) 0.0999-0.1001 Molar

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty