Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol ≥95.5% USP

Supplier: Ricca Chemical
Danger
Synonyms: Ethyl alcohol

R2916000-1A
RCR29160001AEA 149.95 USD InStock
RCR29160001A
Ethanol ≥95.5% USP
Ethanol


Formula: C₂H₅OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay 94.9-96.0 % (v/v)
Acidity or Alkalinity To Pass Test
Clarity To Pass Test
Color To Pass Test
Specific Gravity 0.812-0.816 at 15.56°C
Identification - Ethanol To Pass Test
Nonvolatile Residue To Pass Test
UV Absorption To Pass Test
Volatile Impurities To Pass Test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty