Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylene glycol, Reagent Grade

Supplier: Ricca Chemical
Danger
Synonyms: 1,2-Ethanediol1,2-Dihydroxyethane1,2-Dihydroxy ethaneEthanediolGlycol alcoholDihydroxyethaneEthylene alcoholEthane-1,2-diolMonoethylene glycol2-HydroxyethanolGlycol

R2944500-500C R2944500-20F R2944500-4C
RCR2944500-4CEA 371.87 USD InStock
RCR2944500-4C RCR2944500-20F RCR2944500500C
Ethylene glycol, Reagent Grade
Ethylene glycol


Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: ∼ 197°C
Melting Pt: -12 - -11°C
Density: 1.1135
Flash Pt: 111°C
MDL Number: MFCD00002885
CAS Number: 107-21-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/734375.jpg
Order Now
Specification Test Results
Appearance Colorless viscous liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty