Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Standard Hard Water, 1000 ppm Hardness as CaCO₃, RICCA Chemical Company

Supplier: RICCA Chemical Company
R3486000-20F
RCR3486000-20FEA 439.81 USD InStock
RCR3486000-20F
Standard Hard Water, 1000 ppm Hardness as CaCO₃, RICCA Chemical Company
Standards Environmental Analysis Standards
Assay (vs. EDTA/Eriochrome Black T): 999-1001 ppm Hardness

Appearance; Colorless liquid.

Appearance; Colorless liquiid.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty