Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 20% (v/v)

Supplier: Ricca Chemical
Danger
Synonyms: Chlorohydric acidHydrochlorideHydrogen chloride gasMuriatic acid

R3574000-20F R3574000-10F R3574000-1A R3574000-500A R3574000-4A
RC357432EA 85.56 USD InStock
RC357432 RCR3574000-10F RCR3574000-20F RC357416 RC3574-1
Hydrochloric acid 20% (v/v)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to slightly greenish-yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 19.92-20.08 % (v/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty