Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.0100 N (N/100)

Supplier: Ricca Chemical
Synonyms: Muriatic acidHydrochloric acid solution

R3590000-4A R3590000-500A R3590000-1A R3590000-20F R3590000-120A R3590000-10F
RCR3590000-10FEA 408.13 USD InStock
RCR3590000-10F RCR3590000500A RC359032 RC35901 RC3590-5 RC3590-4
Hydrochloric acid 0.0100 N (N/100)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.00995-0.01005 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty