Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.100 N (N/10)

Supplier: Ricca Chemical
Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

R3600000-1A R3600000-20F R3600000-500A R3600000-10F R3600000-4A R3600000-20E1
RCR3600000-10FEA 158.67 USD InStock
RCR3600000-10F RC3600-16 RC36001 RC36005 RC3600-5HP RC360032
Hydrochloric acid 0.100 N (N/10)
Hydrochloric acid
Standardized at 25 °C against chemicals whose certification is traceable to NIST.


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.0999-0.1001 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty