Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.500 N (N/2)

Supplier: Ricca Chemical
Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

R3650000-10F R3650000-500A R3650000-4A R3650000-20F R3650000-1A
RCR3650000500AEA 37.44 USD InStock
RCR3650000500A RC365032 RC365025 RC36501 RC36505
Hydrochloric acid 0.500 N (N/2)
Hydrochloric acid
Standardized at 25°C against chemicals whose certification is traceable to NIST.


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.4998-0.5002 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty