Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 3.00 N from NF/USP materials

Supplier: Ricca Chemical
Danger
Synonyms: Muriatic acid

R3720100-1A
RCR3720100-1AEA 176.83 USD InStock
RCR3720100-1A
Hydrochloric acid 3.00 N from NF/USP materials
Hydrochloric acid
3.00 Normal Hydrochloric Acid titrant made from NF/USP materials

  • ±0.003 N at 20˚C


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to slightly greenish-yellow liquid
Assay 2.997-3.003 N at 20°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty