Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) acetate 10% (w/v) in aqueous solution with acetic acid (1% (v/v))

Supplier: Ricca Chemical
Synonyms: Acetic acid Lead(II) salt

R4293500-1A
RCR4293500-1AEA 172.52 USD InStock
RCR4293500-1A
Lead(II) acetate 10% (w/v) in aqueous solution with acetic acid (1% (v/v))
Lead (II) acetate


MW: 325.29 g/mol
MDL Number: MFCD00012452
UN: 1616
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty