Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol absolute ACS

Supplier: Ricca Chemical
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

R4829000-20E1 R4829000-25C R4829000-20E4 R4829000-500A R4829000-4X4A
RCR482900020E4EA 333.97 USD InStock
RCR482900020E4 RCR4829000500A RCR48290004X4A RCR4829000-25C RCR482900020E1
Methanol absolute ACS
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: ∼ 64.5°C
Melting Pt: -98°C
Density: 0.7866
Flash Pt: 11°C
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Assay min 99.8 %
Color (APHA) max 10
Solubility in Water To Pass Test
Carbonyl Compounds: Acetaldehyde max 0.001 %
Carbonyl Compounds: Acetone max 0.001 %
Carbonyl Compounds: Formaldehyde max 0.001 %
Water (H₂O) max 0.1 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty