Would you like to log in again?
Mordant black 11 triturate solution R 1% (w/w) in sodium chloride Reag. Ph. Eur.
Supplier: Ricca Chemical
R5215000-100A6
RCR52150-100A6EA 80.95 USD InStock
RCR52150-100A6
Mordant black 11 triturate solution R 1% (w/w) in sodium chloride Reag. Ph. Eur.
Mordant black 11 triturate
Order Now
Specification Test Results
Appearance Gray solid
Suitability for Use Wine Red (excess Ca/Mg) - Pure Blue (excess EDTA)
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty