Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Pyridylazo)resorcinol monosodim salt monohydrate 0.1% (w/v) indicator for complexometry

Supplier: Ricca Chemical
Synonyms: PARPAR monosodium salt monohydrate4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

R5488400-100C
RCR5488400100CEA 83.08 USD InStock
RCR5488400100C
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate 0.1% (w/v) indicator for complexometry
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate


Formula: C₁₁H₈N₃NaO₂·1H₂O
MW: 255.21 g/mol (237.19 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150712
CAS Number: 16593-81-0
Order Now
Specification Test Results
Appearance Dark yellow liquid
Suitability for Use Yellow (excess EDTA) - Pink (excess metal)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty