Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride 100 g/L in purified water Reag. Ph. Eur. for nitrates

Supplier: Ricca Chemical
Synonyms: Potassiumchloride

R5884200-500A R5884200-120A R5884200-250A
RCR5884200250AEA 98.65 USD InStock
RCR5884200250A RCR5884200120A RCR5884200500A
Potassium chloride 100 g/L in purified water Reag. Ph. Eur. for nitrates
Potassium chloride


Formula: KCl
MW: 74.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Silver Nitrate/Potassium Chromate) 99-101 g/L

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty