Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L)

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7260000-120A R7260000-500A R7260000-4A R7260000-20F R7260000-1A
RC726032EA 64.45 USD InStock
RC726032 RC72604 RC7260-1 RC726016 RCR7260000-20F
Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L)
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.96 - 10.04 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty