Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) for Kjeldahl nitrogen analysis nitrogen-free, purified

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7290000-15E R7290000-20F R7290000-55E R7290000-4A R7290000-20E1 R7290000-1A
RC72905EA 580.11 USD InStock
RC72905 RC7290-5HP RCR7290000-15E RCR7290000-55E RC72901 RCR7290000-1A
Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) for Kjeldahl nitrogen analysis nitrogen-free, purified
Sodium hydroxide
Nitrogen-free, suitable for Kjeldahl nitrogen tests.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ~100 °C
Melting Pt: 12 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Hydroxide Alkalinity as NaOH Minimum 49.5% (w/w)
Iron (Fe) 0.0008% max (8 ppm)
Sodium Carbonate Na2CO3 0.4% max
Sodium Chloride (NaCl) 2.20% max
Total Alkalinity as Sodium Oxide (Na2O) Minimum 38.35% (w/w)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty