Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.0200 N (N/50) (1 ml = 1 mg CaCO₃)

Supplier: Ricca Chemical
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

R7300000-20F R7300000-500A R7300000-4A R7300000-1A
RCR7300000500AEA 33.39 USD InStock
RCR7300000500A RCR7300000-20F RC73001 RC730032
Sodium hydroxide 0.0200 N (N/50) (1 ml = 1 mg CaCO₃)
Sodium hydroxide
APHA for acidity.


Formula: NaOH
MW: 40 g/mol
Melting Pt: 0.0°C
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.01995-0.02005 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty