Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 12.5 g/L 0.313 N suitable for crude fiber analysis of food and feed

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7375000-500A R7375000-20F R7375000-1A R7375000-4A
RCR7375000500AEA 31.41 USD InStock
RCR7375000500A RC73751 RCR7375000-1A RC73755
Sodium hydroxide 12.5 g/L 0.313 N suitable for crude fiber analysis of food and feed
Sodium hydroxide
AOAC, for crude fiber.


Formula: NaOH
MW: 40 g/mol
Melting Pt: 0.0°C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: UN1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.3125-0.3135 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty