Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10.0 N

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7470000-20F R7470000-10F R7470000-500A R7470000-4A
RCR7470000-10FEA 337.41 USD InStock
RCR7470000-10F RCR7470000500A RC747001 RC7470-5
Sodium hydroxide 10.0 N
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: Approximately 0°C
Density: 1.33
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: UN1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.95-10.05 N at 20°C
Heavy Metals (as Pb) max 10 ppm
Iron (Fe) max 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty