Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10.0 N , from NF/USP material

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7470050-1A R7470050-120A R7470050-4A
RCR7470050-1AEA 82.35 USD InStock
RCR7470050-1A RCR7470050120A RCR7470050-4A
Sodium hydroxide 10.0 N , from NF/USP material
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: Approximately 0°C
Density: 1.33
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: UN1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.95-10.05 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty