Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium hydroxide 0.100 Normal in methanol

Supplier: Ricca Chemical
Danger

R8384000-4C R8384000-1C
RCR8384000-1CEA 990.02 USD InStock
RCR8384000-1C RCR8384000-4C
Tetrabutylammonium hydroxide 0.100 Normal in methanol
Tetrabutylammonium hydroxide


Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Boiling Pt: ∼ 64.5°C
Density: 0.8000
Flash Pt: ∼ 11°C
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 1230
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to slightly yellow liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 0.0995-0.1005 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty