Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium hydroxide 10% (w/w) in aqueous solution

Supplier: Ricca Chemical
Danger

R8386000-500C
RCR8386000500CEA 347.57 USD InStock
RCR8386000500C
Tetramethylammonium hydroxide 10% (w/w) in aqueous solution
Tetramethylammonium hydroxide


Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: ~102 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
UN: 1835
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to yellow liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 10.0 - 10.2 % (w/w)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty