Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% ACS

Supplier: Ricca Chemical
Danger
Synonyms: Muriatic acidHydrochloric acid solution

RABH0010-55E RABH0010-4C RABH0010-2.5D1
RCRABH0010-4CEA 466.55 USD InStock
RCRABH0010-4C RCRABH0010-55E RCRABH001025D1
Hydrochloric acid 37% ACS
Hydrochloric acid
Testing reagent for ethanol plants.


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Ammonium (NH₄) max 3 ppm
Arsenic (As) max 0.01 ppm
Assay 36.5-38.0 % HCl
Bromide (Br) max 0.005 %
Color (APHA) max 10
Free Chlorine (Cl) max 1 ppm
Heavy Metals (as Pb) max 1 ppm
Iron (Fe) max 0.2 ppm
Residue after Ignition max 5 ppm
Sulfate (SO₄) max 1 ppm
Sulfite (SO₃) max 1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty