Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% for trace metal analysis

Supplier: Ricca Chemical
Danger
Synonyms: Muriatic acid

RABH0020-2.5D
RCRABH0022.5D1EA 145.93 USD InStock
RCRABH0022.5D1
Hydrochloric acid 37% for trace metal analysis
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Aluminum (Al) max 10.0 ppm
Antimony (Sb) max 0.005 ppm
Arsenic (As) max 0.005 ppm
Assay 34.0-38.0%
Boron (B) max 0.1ppm
Bromide (Br) max 0.005%
Calcium (Ca) max 0.3 ppm
Chromium (Cr) max 0.2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty