Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97.0%, pellets ACS

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

RDCS0530-1000B1
RCRDCS05301000EA 139.28 USD InStock
RCRDCS05301000
Sodium hydroxide ≥97.0%, pellets ACS
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay min 97.0 %
Heavy Metals (as Ag) max 0.002 %
Nitrogen Compounds (as N) max 0.001 %
Phosphate (PO₄) max 0.001 %
Sodium Carbonate (Na₂CO₃) max 1.0 %
Sulfate (SO₄) max 0.003 %
Magnesium (Mg) max 0.002 %
Chloride (Cl) max 0.005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty