Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amidosulfonic acid, powder ACS

Supplier: Ricca Chemical
Danger
Synonyms: Sulfamic acidAminosulfonic acid Sulfamidic acid Amidosulfonic acid

RDCS1100-100B1 RDCS1100-500B1 RDCS1100-2.5A5
RCRDCS110100B1EA 94 USD InStock
RCRDCS110100B1 RCRDCS1100500B RCRDCS1102.5A5
Amidosulfonic acid, powder ACS
Amidosulphonic acid


Formula: H₃NO₃S
MW: 97.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
UN: 2967
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg
Order Now
Specification Test Results
Assay (dried basis) 99.3-100.3 %
Heavy Metals (as Pb) max 0.001 %
Sulfate (SO₄) max 0.05 %
Chloride (Cl) max 0.001 %
Iron (Fe) max 5 ppm
Insoluble Matter max 0.01 %
Residue after Ignition max 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty