Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Tartaric acid ACS

Supplier: Ricca Chemical
Danger
Synonyms: L-Tartaric AcidTartaric Acid(2R,3R)-(+)-Tartaric acid(R,R)-(+)-2,3-Dihydroxysuccinic acid(R,R)-(+)-Tartaric acid(2R,3R)-2,3-Dihydroxysuccinic acid

RDCT0030-125B1 RDCT0030-500B1
RCRDCT003500B1EA 157.08 USD InStock
RCRDCT003500B1 RCRDCT003125B1
L(+)-Tartaric acid ACS
L(+)-Tartaric acid


Formula: C₄H₆O₆
MW: 150.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/32/2297232.jpg
Order Now
Specification Test Results
Assay min 99.0 %
Heavy Metals (as Pb) max 5 ppm
Oxalate (C₂O₄) To Pass Test
Phosphate (PO₄) max 0.001 %
Sulfur compounds (as SO₄) max 0.002 %
Chloride (Cl) max 0.001 %
Iron (Fe) max 5 ppm
Insoluble Matter max 0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty