Would you like to log in again?
HydroSpec® Water Standards for Karl Fischer Titration
Supplier: RICCA Chemical Company
HydroSpec®
RK420000-10X5R RK430000-10X5R RK410000-10X5R
RCRK430000105REA 227.3 USD InStock
RCRK430000105R RCRK420000105R RCRK410000105R
HydroSpec® Water Standards for Karl Fischer Titration
Standards Water Determination (by Karl Fischer method) Standards
Achieve high performance and reliable results. Hydrospec® supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.
  • Reliable, Safe, Pyridine free
  • Stable Formulations and Comprehensive Chemical Compatability
  • 5 year shelf life
  • Technical Support Every Step of the Way
  • Tight specifications for More Accurate Reults and Low Lot-to-Lot Variability
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty