Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ACS

Supplier: Ricca Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

RSOC0020-1C RSOC0020-500C RSOC0020-4C
RCRSOC0020-1CEA 94.17 USD InStock
RCRSOC0020-1C RCRSOC0020-4C RCRSOC0020500C
Chloroform ACS
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 60.5 - 61.2°C
Melting Pt: -63.5°C
Density: 1.4840
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Assay min 99.8 %
Color (APHA) max 10
Acetone and Aldehyde To Pass Test
Free Chlorine (Cl) To Pass Test
Lead (Pb) max 0.05 ppm
Acid and Chloride To Pass Test
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Substances Darkened by Sulfuric Acid To Pass Test

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty