Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexane ACS

Supplier: Ricca Chemical
Danger
Synonyms: Hexahydro-BenzeneHexamethylene

RSOC8500-4C
RCRSOC8500-4CEA 266.69 USD InStock
RCRSOC8500-4C
Cyclohexane ACS
Cyclohexane


Formula: C₆H₁₂
MW: 84.16 g/mol
Melting Pt: 6.5°C
Density: 0.7790
Flash Pt: -20°C
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Assay min 99.0 %
Color (APHA) max 10
Water (H₂O) max 0.02 %
Residue after evaporation (Non-Volatile Matter) max 0.002 %
Substances Darkened by Sulfuric Acid To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty