Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 37% in aqueous solution (formalin) ACS

Supplier: Ricca Chemical
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

RSOF0010-4A RSOF0010-500A
RCRSOF0010500AEA 67.94 USD InStock
RCRSOF0010500A RCRSOF0010-4A
Formaldehyde 37% in aqueous solution (formalin) ACS
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III
Order Now
Specification Test Results
Heavy Metals (as Pb) 5 ppm max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty