Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine ACS

Supplier: Ricca Chemical
Warning
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

RSOG0020-4A RSOG0020-1A RSOG0020-500A RSOG0020-20F
RCRSOG0020-4AEA 270.4 USD InStock
RCRSOG0020-4A RCRSOG0020-20F RCRSOG0020-1A RCRSOG0020500A
Glycerine ACS
Glycerine


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 544°C (dec.)
Melting Pt: 18.2°C
Density: 1.2636
Flash Pt: 199°C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Assay min 99.5 %
Color (APHA) max 10
Neutrality To Pass Test
Acrolein and Glucose To Pass Test
Chlorinated compounds (as Cl) max 0.003 %
Heavy Metals (as Pb) max 2 ppm
Sulfate (SO₄) max 0.001 %
Water (H₂O) max 0.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty